Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Boc-amino)-5-fluorobenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 2-(Boc-amino)-5-fluorobenzeneboronic acid pinacol ester
Formula Weight 337.19g/mol
Synonym 2-(Boc-amino)-5-fluorophenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6240803

 
alfa aesar™
H62408-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62408MD

 
alfa aesar™
H62408-MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material 2-(Boc-amino)-5-fluorobenzeneboronic acid pinacol ester
Physical Form Solid
Assay 96%
Formula Weight 337.19g/mol
Synonym 2-(Boc-amino)-5-fluorophenylboronic acid pinacol ester