missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Boc-amino)-5-cyanopyridine, 97%

$228.22 - $909.68

Chemical Identifiers

CAS 902837-44-9
Molecular Formula C11H13N3O2
Molecular Weight (g/mol) 219.244
MDL Number MFCD07781163
InChI Key JDQQNZYKHZVLAM-UHFFFAOYSA-N
Synonym 2-boc-amino pyridine-5-carbonitrile
PubChem CID 42553119
IUPAC Name tert-butyl N-(5-cyanopyridin-2-yl)carbamate
SMILES CC(C)(C)OC(=O)NC1=NC=C(C=C1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5007903 View Documents Alfa Aesar™
H5007903
1g Each for $228.22
 
AAH5007906 View Documents Alfa Aesar™
H5007906
5g Each for $909.68
 
Specifications

Chemical Identifiers

902837-44-9
219.244
JDQQNZYKHZVLAM-UHFFFAOYSA-N
42553119
CC(C)(C)OC(=O)NC1=NC=C(C=C1)C#N
C11H13N3O2
MFCD07781163
2-boc-amino pyridine-5-carbonitrile
tert-butyl N-(5-cyanopyridin-2-yl)carbamate

Specifications

170°C to 175°C
902837-44-9
MFCD07781163
JDQQNZYKHZVLAM-UHFFFAOYSA-N
tert-butyl N-(5-cyanopyridin-2-yl)carbamate
42553119
97%
1g
C11H13N3O2
2-boc-amino pyridine-5-carbonitrile
CC(C)(C)OC(=O)NC1=NC=C(C=C1)C#N
219.244
219.24
2-(Boc-amino)-5-cyanopyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.

P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c

H302-H319

TSCA : No

Recommended Storage : Ambient temperatures