missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Boc-amino)-5-bromothiazole, 97%

Manufacturer:  Alfa Aesar™ H6182803

 View more versions of this product

Catalog No. AAH6182803



Description

Description

Applications
2-(Boc-amino)-5-bromothiazole is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

2-(Boc-amino)-5-bromothiazole
405939-39-1
MFCD07368614
OIBKBVFFZYCBAQ-UHFFFAOYSA-N
tert-butyl N-(5-bromo-1,3-thiazol-2-yl)carbamate
21300388
Solid
1g
C8H11BrN2O2S
5-bromo-thiazol-2-yl-carbamic acid tert-butyl ester
CC(C)(C)OC(=O)NC1=NC=C(S1)Br
279.152
279.15
97%

Chemical Identifiers

405939-39-1
279.152
OIBKBVFFZYCBAQ-UHFFFAOYSA-N
21300388
CC(C)(C)OC(=O)NC1=NC=C(S1)Br
C8H11BrN2O2S
MFCD07368614
5-bromo-thiazol-2-yl-carbamic acid tert-butyl ester
tert-butyl N-(5-bromo-1,3-thiazol-2-yl)carbamate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents