missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Boc-amino)-4-cyanopyridine, 97%

$103.76 - $389.43

Chemical Identifiers

CAS 737000-78-1
Molecular Formula C11H13N3O2
Formula Weight 219.24
MDL Number MFCD03791311
Synonym 2-(Boc-amino)isonicotinonitrile; 2-(tert-Butoxycarbonylamino)-4-cyanopyridine
Chemical Name or Material 2-(Boc-amino)-4-cyanopyridine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1984803 SDS Alfa Aesar™
L1984803
1g Each for $103.76
AAL1984806 SDS Alfa Aesar™
L1984806
5g Each for $389.43
Specifications Description & Specifications

Chemical Identifiers

737000-78-1
219.24
2-(Boc-amino)isonicotinonitrile; 2-(tert-Butoxycarbonylamino)-4-cyanopyridine
C11H13N3O2
MFCD03791311
2-(Boc-amino)-4-cyanopyridine

Specifications

737000-78-1
≈175°C
MFCD03791311
2-(Boc-amino)isonicotinonitrile; 2-(tert-Butoxycarbonylamino)-4-cyanopyridine
97%
C11H13N3O2
219.24
1g
2-(Boc-amino)-4-cyanopyridine

Safety and Handling

GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
P280f-P305+P351+P338
Wear protective clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning