missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[Bis(methylthio)methylene]malononitrile, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H5350306

 View more versions of this product

Catalog No. AAH5350306



Description

Description

Applications
2-[Bis(methylthio)methylene]malononitrile is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications

Specifications

2-[Bis(methylthio)methylene]malononitrile
5g
C6H6N2S2
MFCD00052730
2-bis methylsulfanyl methylidene propanedinitrile
CSC(=C(C#N)C#N)SC
170.248
170.25
79°C to 82°C
5147-80-8
(NC)2C=C(SCH3)2
UN3439
FICQFRCPSFCFBY-UHFFFAOYSA-N
2-[bis(methylsulfanyl)methylidene]propanedinitrile
99229
97%

Chemical Identifiers

5147-80-8
170.248
FICQFRCPSFCFBY-UHFFFAOYSA-N
99229
CSC(=C(C#N)C#N)SC
C6H6N2S2
MFCD00052730
2-bis methylsulfanyl methylidene propanedinitrile
2-[bis(methylsulfanyl)methylidene]propanedinitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c

H301+H311+H331

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents