Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-[Bis(3,5-dimethylphenyl)phosphino]benzaldehyde, 97%

Chemical Identifiers

Chemical Name or Material 2-[Bis(3,5-dimethylphenyl)phosphino]benzaldehyde
CAS 669091-00-3
Formula Weight 346.4
MDL Number MFCD15143662
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH63117MC

  SDS
alfa aesar™
H63117MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63117ME

  SDS
alfa aesar™
H63117ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 346.4
Melting Point 106° to 110°C
MDL Number MFCD15143662
Physical Form Solid
Color Yellow
Quantity 100mg
Sensitivity Air Sensitive
Chemical Name or Material 2-[Bis(3,5-dimethylphenyl)phosphino]benzaldehyde
CAS 669091-00-3
Assay 97%