missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[Bis(3,5-dimethylphenyl)phosphino]benzaldehyde, 97%, Alfa Aesar™

Manufacturer:  Alfa Aesar H63117MC

 View more versions of this product

Catalog No. AAH63117MC



Specifications

Specifications

2-[Bis(3,5-dimethylphenyl)phosphino]benzaldehyde
Yellow
100mg
C23H23OP
2-bis 3,5-dimethylphenyl phosphino benzaldehyde, 2-bis 3,5-dimethylphenyl phosphanyl benzaldehyde, 2-bis 3,5-dimethylphenyl phosphino benzaldehyde, 2-bis 3,5-dimethylphenyl phosphino benzaldehyde, acmc-20ao6m, 2-bis 3,5-dimethylphenyl phosphino benzaldehyde,min
CC1=CC(=CC(=C1)P(C2=CC=CC=C2C=O)C3=CC(=CC(=C3)C)C)C
346.41
346.4
97%
Air Sensitive
106°C to 110°C
669091-00-3
MFCD15143662
PQIAXRGPKDEYLW-UHFFFAOYSA-N
2-bis(3,5-dimethylphenyl)phosphanylbenzaldehyde
71310677
Solid

Chemical Identifiers

669091-00-3
346.41
PQIAXRGPKDEYLW-UHFFFAOYSA-N
71310677
CC1=CC(=CC(=C1)P(C2=CC=CC=C2C=O)C3=CC(=CC(=C3)C)C)C
C23H23OP
MFCD15143662
2-bis 3,5-dimethylphenyl phosphino benzaldehyde, 2-bis 3,5-dimethylphenyl phosphanyl benzaldehyde, 2-bis 3,5-dimethylphenyl phosphino benzaldehyde, 2-bis 3,5-dimethylphenyl phosphino benzaldehyde, acmc-20ao6m, 2-bis 3,5-dimethylphenyl phosphino benzaldehyde,min
2-bis(3,5-dimethylphenyl)phosphanylbenzaldehyde
SDS Documents