Please login to your online account to display your discounted pricing

2-(Bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino)benzaldehyde, 97%, ACROS Organics™


Quantity 100mg

Manufacturer: thermo fisher scientific  AC435131000

 View more versions of this product

Catalog No. AC435131000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Powder
Formula Weight 574.78
Quantity 100mg
Color Yellow
Melting Point 122° to 126°C
CAS Min % 96.0
CAS Max % 100.0
Elemental Analysis 74.22 to 80.41% C
NMR (1H-NMR) Consistent with structure, (31P-NMR) Consistent with structure, NMR (19F-NMR), Consistent with structure
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay Percent Range 97%
CAS 1202865-21-1
MDL Number MFCD15143663
Chemical Name or Material 2-(Bis(3, 5-di-tert-Butyl-4-methoxyphenyl)phosphino)benzaldehyde
Molecular Formula C37H51O3P
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-(Bis(3, 5-di-tert-Butyl-4-methoxyphenyl)phosphino)benzaldehyde
CAS 1202865-21-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C37H51O3P
Formula Weight 574.78
MDL Number MFCD15143663
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents