Please login to your online account to display your discounted pricing

2-(Bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino)benzaldehyde, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-(Bis(3, 5-di-tert-Butyl-4-methoxyphenyl)phosphino)benzaldehyde
CAS 1202865-21-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C37H51O3P
Formula Weight 574.78
MDL Number MFCD15143663
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC435132500

  SDS
thermo fisher scientific
AC435132500
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 574.78
Physical Form Powder
Assay ≥96.0%
Quantity 250mg
Color Yellow
Melting Point 122° to 126°C
CAS Max % 100.0
CAS Min % 96.0
Elemental Analysis 74.22 to 80.41% C
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 1202865-21-1
Chemical Name or Material 2-(Bis(3, 5-di-tert-Butyl-4-methoxyphenyl)phosphino)benzaldehyde
MDL Number MFCD15143663
Assay Percent Range 97%
Molecular Formula C37H51O3P
Infrared Spectrum Authentic
NMR (1H-NMR) Consistent with structure, (31P-NMR) Consistent with structure, NMR (19F-NMR), Consistent with structure
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.