missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[Bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]benzaldehyde, 97%, Alfa Aesar™

Manufacturer:  Alfa Aesar H63980MC

 View more versions of this product

Catalog No. AAH63980MC



Specifications

Specifications

2-[Bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]benzaldehyde
Yellow
100mg
C37H51O3P
2-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphino benzaldehyde, 2-bis 3,5-di-t-butyl-4-methoxyphenyl phosphino benzaldehyde, 2-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphanyl benzaldehyde, acmc-20ao6l, 2-bis 3,5-di-t-butyl-4-methoxyphenyl phosphino benzaldehyde, min, 2-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphino benzaldehyde
CC(C)(C)C1=CC(=CC(=C1OC)C(C)(C)C)P(C2=CC=CC=C2C=O)C3=CC(=C(C(=C3)C(C)(C)C)OC)C(C)(C)C
574.786
574.77
97%
Air Sensitive
120°C to 125°C
1202865-21-1
MFCD15143663
PAWZHFURVJBYGV-UHFFFAOYSA-N
2-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphanylbenzaldehyde
71310909
Solid

Chemical Identifiers

1202865-21-1
574.786
PAWZHFURVJBYGV-UHFFFAOYSA-N
71310909
CC(C)(C)C1=CC(=CC(=C1OC)C(C)(C)C)P(C2=CC=CC=C2C=O)C3=CC(=C(C(=C3)C(C)(C)C)OC)C(C)(C)C
C37H51O3P
MFCD15143663
2-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphino benzaldehyde, 2-bis 3,5-di-t-butyl-4-methoxyphenyl phosphino benzaldehyde, 2-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphanyl benzaldehyde, acmc-20ao6l, 2-bis 3,5-di-t-butyl-4-methoxyphenyl phosphino benzaldehyde, min, 2-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphino benzaldehyde
2-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphanylbenzaldehyde
Documents