missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Biphenylyl Glycidyl Ether, TCI America™

$36.67 - $366.67

Chemical Identifiers

CAS 7144-65-2
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.275
MDL Number MFCD00046989
InChI Key DNVXWIINBUTFEP-UHFFFAOYSA-N
Synonym 2-biphenylyl glycidyl ether, opp-g, 3-2-xenyloxy propylene oxide, 2-1,1'-biphenyl-2-yloxy methyl oxirane, ccris 2065, 3-2-biphenyloxy propylene oxide, 3-2-xenolyl-1,2-epoxypropane, biphenyl-2-yl 2,3-epoxypropyl ether, ether, 2-biphenylyl 2,3-epoxypropyl, oxirane, 2-1,1'-biphenyl-2-yloxy methyl
PubChem CID 97875
IUPAC Name 2-[(2-phenylphenoxy)methyl]oxirane
SMILES C1C(O1)COC2=CC=CC=C2C3=CC=CC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B560125G View Documents TCI America
B560125G
25g Each for $36.67
 
B5601500G View Documents TCI America
B5601500G
500g Each for $366.67
 
Specifications

Chemical Identifiers

7144-65-2
226.275
DNVXWIINBUTFEP-UHFFFAOYSA-N
97875
C1C(O1)COC2=CC=CC=C2C3=CC=CC=C3
C15H14O2
MFCD00046989
2-biphenylyl glycidyl ether, opp-g, 3-2-xenyloxy propylene oxide, 2-1,1'-biphenyl-2-yloxy methyl oxirane, ccris 2065, 3-2-biphenyloxy propylene oxide, 3-2-xenolyl-1,2-epoxypropane, biphenyl-2-yl 2,3-epoxypropyl ether, ether, 2-biphenylyl 2,3-epoxypropyl, oxirane, 2-1,1'-biphenyl-2-yloxy methyl
2-[(2-phenylphenoxy)methyl]oxirane

Specifications

200°C
32°C
7144-65-2
MFCD00046989
DNVXWIINBUTFEP-UHFFFAOYSA-N
2-[(2-phenylphenoxy)methyl]oxirane
97875
Crystal-Lump at 20°C
2-Biphenylyl Glycidyl Ether
White
25g
C15H14O2
2-biphenylyl glycidyl ether, opp-g, 3-2-xenyloxy propylene oxide, 2-1,1'-biphenyl-2-yloxy methyl oxirane, ccris 2065, 3-2-biphenyloxy propylene oxide, 3-2-xenolyl-1,2-epoxypropane, biphenyl-2-yl 2,3-epoxypropyl ether, ether, 2-biphenylyl 2,3-epoxypropyl, oxirane, 2-1,1'-biphenyl-2-yloxy methyl
C1C(O1)COC2=CC=CC=C2C3=CC=CC=C3
226.275
226.28
≥97.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0062

RTECSNumber : RR0356000

TSCA : No

Recommended Storage : Refrigerator