Please login to your online account to display your discounted pricing

2-Biphenylyl Glycidyl Ether 95%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2-Biphenylyl glycidyl ether
Name Note 95%
CAS 7144-65-2
CAS Min % 94.0
CAS Max % 100.0
Molecular Formula C15H14O2
Formula Weight 226.27
MDL Number MFCD00046989
Synonym 3-(2-Biphenylyloxy)-1, 2-epoxypropane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC188170050

  SDS
thermo fisher scientific
188170050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 226.27
Quantity 5g
TSCA TSCA
RTECS Number RR0356000
EINECS Number 230-451-0
Beilstein 17, IV, 995
CAS Max % 100.0
CAS Min % 94.0
Synonym 3-(2-Biphenylyloxy)-1, 2-epoxypropane
Chemical Name or Material 2-Biphenylyl glycidyl ether
Name Note 95%
CAS 7144-65-2
Molecular Formula C15H14O2
MDL Number MFCD00046989