missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bibenzylcarboxylic acid, 98%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 348810250

 View more versions of this product

Catalog No. AC348810250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
226.27
White to off-white
100.0
Authentic
C15H14O2
97.5
2-Bibenzylcarboxylic acid
2-Phenethylbenzoic acid
Crystalline Powder
25g
127° to 132°C
4890-85-1
C6H5(CH2)2C6H4CO2H
98%
MFCD00002485
Glass bottle

Chemical Identifiers

4890-85-1
100.0
C15H14O2
226.27
2-Phenethylbenzoic acid
≥97.5%
97.5
98%
C6H5(CH2)2C6H4CO2H
MFCD00002485
2-Bibenzylcarboxylic acid

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 225-511-8

TSCA : TSCA