Please login to your online account to display your discounted pricing

2-Bibenzylcarboxylic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Bibenzylcarboxylic acid
CAS 4890-85-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C15H14O2
Linear Formula C6H5(CH2)2C6H4CO2H
Formula Weight 226.27
MDL Number MFCD00002485
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC348810250

  SDS
fisher scientific acros
348810250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC348811000

  SDS
thermo fisher scientific
348811000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 226.27
Quantity 25g
Color White to off-white
TSCA TSCA
Melting Point 127° to 132°C
EINECS Number 225-511-8
CAS Max % 100.0
CAS 4890-85-1
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Linear Formula C6H5(CH2)2C6H4CO2H
Molecular Formula C15H14O2
Assay Percent Range 98%
CAS Min % 97.5
MDL Number MFCD00002485
Chemical Name or Material 2-Bibenzylcarboxylic acid
Packaging Glass bottle
Synonym 2-Phenethylbenzoic acid
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.