missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Benzylthio)ethanol, TCI America™

$87.08 - $268.89

Chemical Identifiers

CAS 3878-41-9
Molecular Formula C9H12OS
Molecular Weight (g/mol) 168.254
MDL Number MFCD00014039
InChI Key ANMHSIXPUAKNLM-UHFFFAOYSA-N
Synonym 2-Hydroxyethyl Benzyl Sulfide
PubChem CID 77486
IUPAC Name 2-benzylsulfanylethanol
SMILES C1=CC=C(C=C1)CSCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B40875G View Documents TCI America
B40875G
5g Each for $87.08
 
B408725G View Documents TCI America
B408725G
25g Each for $268.89
 
Specifications

Chemical Identifiers

3878-41-9
168.254
ANMHSIXPUAKNLM-UHFFFAOYSA-N
77486
C1=CC=C(C=C1)CSCCO
C9H12OS
MFCD00014039
2-Hydroxyethyl Benzyl Sulfide
2-benzylsulfanylethanol

Specifications

120°C
5g
C9H12OS
2-Hydroxyethyl Benzyl Sulfide
C1=CC=C(C=C1)CSCCO
168.254
168.25
≥98.0% (GC)
Yellow
3878-41-9
MFCD00014039
ANMHSIXPUAKNLM-UHFFFAOYSA-N
2-benzylsulfanylethanol
77486
Clear Liquid at 20°C
2-(Benzylthio)ethanol
Safety and Handling

Safety and Handling

TSCA : No