Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Benzylthiazolidine-4-carboxylic acid, 97%

Chemical Identifiers

Chemical Name or Material 2-Benzylthiazolidine-4-carboxylic acid
CAS 50739-30-5
Formula Weight 223.29
MDL Number MFCD09936775
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6220903

 
alfa aesar™
H6220903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6220906

 
alfa aesar™
H6220906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62209MD

 
alfa aesar™
H62209MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 223.29
Chemical Name or Material 2-Benzylthiazolidine-4-carboxylic acid
Physical Form Solid
Assay 97%
MDL Number MFCD09936775
CAS 50739-30-5
Quantity 1g