missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Benzylthiazolidine-4-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H6220906

 View more versions of this product

Catalog No. AAH6220906



Specifications

Specifications

2-Benzylthiazolidine-4-carboxylic acid
50739-30-5
MFCD09936775
SNFSDSLKMQQXDU-UHFFFAOYSA-N
2-benzyl-1,3-thiazolidine-4-carboxylic acid
4285155
Solid
5g
C11H13NO2S
2-benzylthiazolidine-4-carboxylic acid, 2-benzyl-thiazolidine-4-carboxylic acid, 2-benzyl-4-thiazolidinecarboxylic acid
C1C(NC(S1)CC2=CC=CC=C2)C(=O)O
223.29
223.29
97%

Chemical Identifiers

50739-30-5
223.29
SNFSDSLKMQQXDU-UHFFFAOYSA-N
4285155
C1C(NC(S1)CC2=CC=CC=C2)C(=O)O
C11H13NO2S
MFCD09936775
2-benzylthiazolidine-4-carboxylic acid, 2-benzyl-thiazolidine-4-carboxylic acid, 2-benzyl-4-thiazolidinecarboxylic acid
2-benzyl-1,3-thiazolidine-4-carboxylic acid
Documents