Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Benzylphenol, 98+%

$48.64 - $544.91

Chemical Identifiers

Chemical Name or Material 2-Benzylphenol
CAS 28994-41-4
Molecular Formula C13H12O
Formula Weight 184.24
MDL Number MFCD00002242
Beilstein 2045725
Merck Index 14,1142
Synonym 2-Hydroxydiphenylmethane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2224609

 
alfa aesar™
B2224609
10g Each for $48.64

AAB2224618

 
alfa aesar™
B2224618
50g Each for $142.24

AAB2224630

 
alfa aesar™
B2224630
250g Each for $544.91
Description & Specifications

Specifications

Formula Weight 184.24
Chemical Name or Material 2-Benzylphenol
TSCA TSCA
Assay ≥98%
Molecular Formula C13H12O
Melting Point 50° to 54°C
MDL Number MFCD00002242
CAS 28994-41-4
Beilstein 2045725
Boiling Point 310° to 312°C
Merck Index 14,1142
Quantity 10g
Flash Point >110°C (230°F)
Synonym 2-Hydroxydiphenylmethane
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.