missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Benzyloxypyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

$202.92 - $552.33

Chemical Identifiers

CAS 1073371-81-9
Molecular Formula C18H22BNO3
Molecular Weight (g/mol) 311.19
MDL Number MFCD06798265
InChI Key IFMSDMPTVPMGOP-UHFFFAOYSA-N
Synonym 2-benzyloxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-benzyloxypyridine-3-boronic acid pinacol ester, acmc-20aajr, 2-benzyloxy pyridin-3-yl boronic acid pinacol ester, 2-benzyloxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 2-benzyloxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-benzoxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-phenylmethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine
PubChem CID 44755162
IUPAC Name 2-phenylmethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=C(OCC2=CC=CC=C2)N=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH26928MD
View Documents Promotion Details
Thermo Scientific Chemicals
H26928MD
250 mg
Each for $202.92
 
AAH2692803
View Documents Promotion Details
Thermo Scientific Chemicals
H2692803
1 g
Each for $552.33
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1073371-81-9
311.19
IFMSDMPTVPMGOP-UHFFFAOYSA-N
44755162
CC1(C)OB(OC1(C)C)C1=C(OCC2=CC=CC=C2)N=CC=C1
C18H22BNO3
MFCD06798265
2-benzyloxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-benzyloxypyridine-3-boronic acid pinacol ester, acmc-20aajr, 2-benzyloxy pyridin-3-yl boronic acid pinacol ester, 2-benzyloxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 2-benzyloxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-benzoxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-phenylmethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine
2-phenylmethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Specifications

1073371-81-9
MFCD06798265
2-benzyloxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-benzyloxypyridine-3-boronic acid pinacol ester, acmc-20aajr, 2-benzyloxy pyridin-3-yl boronic acid pinacol ester, 2-benzyloxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 2-benzyloxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-benzoxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-phenylmethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine
CC1(C)OB(OC1(C)C)C1=C(OCC2=CC=CC=C2)N=CC=C1
311.19
311.19
2-Benzyloxypyridine-3-boronic acid pinacol ester
C18H22BNO3
250 mg
IFMSDMPTVPMGOP-UHFFFAOYSA-N
2-phenylmethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
44755162
95%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only