Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Benzyloxyphenylacetonitrile, 98%

$73.26 - $197.03

Chemical Identifiers

Chemical Name or Material 2-Benzyloxyphenylacetonitrile
CAS 92552-22-2
Molecular Formula C15H13NO
Formula Weight 223.28
MDL Number MFCD00016389
Beilstein 2731076
Synonym 2-Benzyloxybenzyl cyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1924004

 
alfa aesar™
L1924004
2g Each for $197.03

AAL19240ME

 
alfa aesar™
L19240ME
500mg Each for $73.26
Description & Specifications

Specifications

Formula Weight 223.28
Molecular Formula C15H13NO
CAS 92552-22-2
Chemical Name or Material 2-Benzyloxyphenylacetonitrile
Assay 98%
MDL Number MFCD00016389
Beilstein 2731076
Synonym 2-Benzyloxybenzyl cyanide
Melting Point 75° to 78°C
Health Hazard 1 Warning
Quantity 2g
Health Hazard 2 GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P280h
Wear protective gloves/clothing.
Safety and Handling

Warning

GHS P Statement
P280h
Wear protective gloves/clothing.

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.