Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Benzyloxyphenylacetic acid, 98+%

$87.43 - $1,397.66

Chemical Identifiers

Chemical Name or Material 2-Benzyloxyphenylacetic acid
CAS 22047-88-7
Molecular Formula C15H14O3
Formula Weight 242.28
MDL Number MFCD00016822
Beilstein 2653977
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1523703

 
alfa aesar™
A1523703
1g Each for $87.43

AAA1523706

 
alfa aesar™
A1523706
5g Each for $326.32

AAA1523714

 
alfa aesar™
A1523714
25g Each for $1,397.66
Description & Specifications

Specifications

Formula Weight 242.28
Chemical Name or Material 2-Benzyloxyphenylacetic acid
Assay ≥98%
Molecular Formula C15H14O3
MDL Number MFCD00016822
CAS 22047-88-7
Beilstein 2653977
Melting Point 96° to 98°C
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.