missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Benzyloxyphenylacetic acid, 98+%

Manufacturer:  Alfa Aesar™ A1523714

 View more versions of this product

Catalog No. AAA1523714



Specifications

Specifications

2-Benzyloxyphenylacetic acid
25g
C15H14O3
2653977
VFYKRBZHJFJOGQ-UHFFFAOYSA-N
2-(2-phenylmethoxyphenyl)acetic acid
519916
≥98%
96°C to 98°C
22047-88-7
MFCD00016822
2-benzyloxyphenylacetic acid, o-benzyloxyphenylacetic acid, 2-2-benzyloxy phenyl acetic acid, 2-benzyloxy phenyl acetic acid, 2-benzyloxyphenyl acetic acid, benzeneacetic acid,2-phenylmethoxy, 2-2-benzyloxy phenyl acetic acid, 2-2-phenylmethoxy phenyl acetic acid, acmc-20ao3j, 2-benzyloxyphenyl acetic acid
C1=CC=C(C=C1)COC2=CC=CC=C2CC(=O)O
242.274
242.28

Chemical Identifiers

22047-88-7
242.274
VFYKRBZHJFJOGQ-UHFFFAOYSA-N
519916
C1=CC=C(C=C1)COC2=CC=CC=C2CC(=O)O
C15H14O3
MFCD00016822
2-benzyloxyphenylacetic acid, o-benzyloxyphenylacetic acid, 2-2-benzyloxy phenyl acetic acid, 2-benzyloxy phenyl acetic acid, 2-benzyloxyphenyl acetic acid, benzeneacetic acid,2-phenylmethoxy, 2-2-benzyloxy phenyl acetic acid, 2-2-phenylmethoxy phenyl acetic acid, acmc-20ao3j, 2-benzyloxyphenyl acetic acid
2-(2-phenylmethoxyphenyl)acetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents