missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Benzyloxybenzyl Alcohol, TCI America™

$207.78 - $713.48

Chemical Identifiers

CAS 3381-87-1
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.264
InChI Key XRZMRABYCSOXIZ-UHFFFAOYSA-N
PubChem CID 18811
IUPAC Name (2-phenylmethoxyphenyl)methanol
SMILES C1=CC=C(C=C1)COC2=CC=CC=C2CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B29435G View Documents TCI America
B29435G
5g Each for $207.78
 
B294325G View Documents TCI America
B294325G
25g Each for $713.48
 
Specifications

Chemical Identifiers

3381-87-1
214.264
18811
C1=CC=C(C=C1)COC2=CC=CC=C2CO
C14H14O2
XRZMRABYCSOXIZ-UHFFFAOYSA-N
(2-phenylmethoxyphenyl)methanol

Specifications

155°C
5g
C14H14O2
C1=CC=C(C=C1)COC2=CC=CC=C2CO
214.264
214.26
≥97.0% (GC)
White-Yellow
3381-87-1
XRZMRABYCSOXIZ-UHFFFAOYSA-N
(2-phenylmethoxyphenyl)methanol
18811
Crystal at 20°C
2-Benzyloxybenzyl Alcohol
Safety and Handling

Safety and Handling

RTECSNumber : DN8582000

TSCA : No