Please login to your online account to display your discounted pricing

2-Benzyloxybenzaldehyde 98%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  392430500

 View more versions of this product

Catalog No. AC392430500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5980 to 1.6020 (20°C, 589nm)
Density 1.15g/mL
Color Colorless to yellow
Quantity 50g
Chemical Name or Material 2-Benzyloxybenzaldehyde
CAS 5896-17-3
CAS Min % 97.5
CAS Max % 100.0
Physical Form Colorless to yellow liquid
Assay 97.5 % min. (GC)
Assay Percent Range 98%
Molecular Formula C14H12O2
Linear Formula C6H5CH2OC6H4C(=O)H
Formula Weight 212.25g/mol
MDL Number MFCD00016583
Beilstein 08, III, 145

Specifications

Chemical Name or Material 2-Benzyloxybenzaldehyde
CAS 5896-17-3
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5 % min. (GC)
Assay Percent Range 98%
Molecular Formula C14H12O2
Linear Formula C6H5CH2OC6H4C(=O)H
Formula Weight 212.25g/mol
MDL Number MFCD00016583