Please login to your online account to display your discounted pricing

2-Benzyloxybenzaldehyde 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Benzyloxybenzaldehyde
CAS 5896-17-3
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5 % min. (GC)
Assay Percent Range 98%
Molecular Formula C14H12O2
Linear Formula C6H5CH2OC6H4C(=O)H
Formula Weight 212.25g/mol
MDL Number MFCD00016583
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC392430100

  SDS
thermo fisher scientific
392430100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC392430500

  SDS
thermo fisher scientific
392430500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5980 to 1.6020 (20°C, 589nm)
Density 1.15g/mL
Color Colorless to yellow
Quantity 10g
Chemical Name or Material 2-Benzyloxybenzaldehyde
CAS 5896-17-3
CAS Min % 97.5
CAS Max % 100.0
Physical Form Colorless to yellow liquid
Assay 97.5 % min. (GC)
Assay Percent Range 98%
Molecular Formula C14H12O2
Linear Formula C6H5CH2OC6H4C(=O)H
Formula Weight 212.25g/mol
MDL Number MFCD00016583
Beilstein 08, III, 145