missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Benzyloxybenzaldehyde, 98%

$44.14 - $130.89

Chemical Identifiers

CAS 5896-17-3
Molecular Formula C14H12O2
Formula Weight 212.25
MDL Number MFCD00016583
Beilstein 780307
Chemical Name or Material 2-Benzyloxybenzaldehyde
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1286609 SDS Alfa Aesar™
L1286609
10g Each for $44.14
AAL1286618 SDS Alfa Aesar™
L1286618
50g Each for $130.89
Specifications

Description

Applications
2-Benzyloxybenzaldehyde is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

5896-17-3
212.25
780307
C14H12O2
MFCD00016583
2-Benzyloxybenzaldehyde

Specifications

5896-17-3
212.25
780307
10g
98%
C14H12O2
MFCD00016583
46° to 47°C
2-Benzyloxybenzaldehyde