missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Benzyloxybenzaldehyde, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1286618
Description
2-Benzyloxybenzaldehyde is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Benzyloxybenzaldehyde is used as an organic chemical synthesis intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
2-Benzyloxybenzaldehyde | |
46°C to 47°C | |
MFCD00016583 | |
780307 | |
2-benzyloxy benzaldehyde, 2-benzyloxybenzaldehyde, benzaldehyde, 2-phenylmethoxy, o-benzyloxy benzaldehyde, benzaldehyde, o-benzyloxy, o-benzylsalicylaldehyde, 2-benzyl-oxy benzaldehyde, 2-phenylmethoxy benzaldehyde, benzyloxybenzaldehyde, o-benzyloxybenzaldehyde | |
PBEJTRAJWCNHRS-UHFFFAOYSA-N | |
2-phenylmethoxybenzaldehyde | |
344784 | |
98% |
5896-17-3 | |
C14H12O2 | |
50 g | |
Air Sensitive | |
Insoluble in water. | |
O=CC1=CC=CC=C1OCC1=CC=CC=C1 | |
212.25 | |
212.25 |
Chemical Identifiers
5896-17-3 | |
212.25 | |
PBEJTRAJWCNHRS-UHFFFAOYSA-N | |
344784 | |
O=CC1=CC=CC=C1OCC1=CC=CC=C1 |
C14H12O2 | |
MFCD00016583 | |
2-benzyloxy benzaldehyde, 2-benzyloxybenzaldehyde, benzaldehyde, 2-phenylmethoxy, o-benzyloxy benzaldehyde, benzaldehyde, o-benzyloxy, o-benzylsalicylaldehyde, 2-benzyl-oxy benzaldehyde, 2-phenylmethoxy benzaldehyde, benzyloxybenzaldehyde, o-benzyloxybenzaldehyde | |
2-phenylmethoxybenzaldehyde |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only