missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Benzyloxyaniline, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC398300010

 View more versions of this product

Catalog No. AC398300010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Benzyloxyaniline
96.0
Authentic
Crystalline Powder or Lms
97%
C6H5CH2OC6H4NH2
35° to 39°C
White to gray to brown
20012-63-9
100.0
Glass bottle
≥96.0% (GC)
C13H13NO
199.25
MFCD03093873
1g

Chemical Identifiers

20012-63-9
100.0
C13H13NO
199.25
2-Benzyloxyaniline
96.0
97%
C6H5CH2OC6H4NH2
MFCD03093873
≥96.0% (GC)

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning