Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Benzyloxyaniline, 97%

$45.72 - $136.85

Chemical Identifiers

Chemical Name or Material 2-Benzyloxyaniline
CAS 20012-63-9
Molecular Formula C13H13NO
Formula Weight 199.25
MDL Number MFCD03093873
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2706506

  SDS
alfa aesar™
H2706506
5g Each for $136.85

AAH2706503

  SDS
alfa aesar™
H2706503
1g Each for $45.72
Description & Specifications

Specifications

Formula Weight 199.25
Chemical Name or Material 2-Benzyloxyaniline
Assay 97%
Molecular Formula C13H13NO
Melting Point 35° to 39°C
MDL Number MFCD03093873
CAS 20012-63-9
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.