Please login to your online account to display your discounted pricing

2-(Benzyloxy)phenol, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-(Benzyloxy)phenol
CAS 6272-38-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥95.0%
Assay Percent Range 97%
Molecular Formula C13H12O2
Linear Formula C6H5CH2OC6H4OH
Formula Weight 200.24
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC179320100

  SDS
thermo fisher scientific
AC179320100
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC179320050

  SDS
thermo fisher scientific
AC179320050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95.0%
Formula Weight 200.24
Physical Form Liquid or Low Melting
Quantity 10g
Color Colorless to pale yellow
Density 1.1400g/mL
CAS Min % 96.0
CAS Max % 100.0
Linear Formula C6H5CH2OC6H4OH
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Causes serious eye irritation.
Molecular Formula C13H12O2
CAS 6272-38-4
Assay Percent Range 97%
Chemical Name or Material 2-(Benzyloxy)phenol
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Causes serious eye irritation.