Please login to your online account to display your discounted pricing

2-(Benzyloxy)phenol, 97%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC179320050

 View more versions of this product

Catalog No. AC179320050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid or Low Melting
Assay ≥95.0%
Formula Weight 200.24
Quantity 5g
Color Colorless to pale yellow
Density 1.1400g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2-(Benzyloxy)phenol
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C13H12O2
Linear Formula C6H5CH2OC6H4OH
CAS 6272-38-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-(Benzyloxy)phenol
CAS 6272-38-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥95.0%
Assay Percent Range 97%
Molecular Formula C13H12O2
Linear Formula C6H5CH2OC6H4OH
Formula Weight 200.24
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Causes serious eye irritation.

SDS & Safety Documents