Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Benzyloxy-5-hydroxypyridine, 95%

$342.36 - $342.36

Chemical Identifiers

Chemical Name or Material 2-Benzyloxy-5-hydroxypyridine
CAS 725256-57-5
Molecular Formula C12H11NO2
Formula Weight 201.22g/mol
MDL Number MFCD06798281
Synonym 6-Benzyloxy-3-pyridinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2746703

 
alfa aesar™
H27467-03
1g Each for $342.36
Description & Specifications

Specifications

Melting Point 90° to 93°C
Quantity 1g
Chemical Name or Material 2-Benzyloxy-5-hydroxypyridine
CAS 725256-57-5
Assay 95%
Molecular Formula C12H11NO2
Formula Weight 201.22g/mol
MDL Number MFCD06798281
Synonym 6-Benzyloxy-3-pyridinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.