missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Benzyloxy-5-bromopyrimidine, 95%

$112.99 - $253.99

Chemical Identifiers

CAS 742058-39-5
Molecular Formula C11H9BrN2O
Formula Weight 265.11
MDL Number MFCD07375140
Chemical Name or Material 2-Benzyloxy-5-bromopyrimidine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2764803 SDS Alfa Aesar™
H2764803
1g Each for $253.99
AAH27648MD SDS Alfa Aesar™
H27648MD
250mg Each for $112.99
Specifications Description & Specifications

Chemical Identifiers

742058-39-5
265.11
2-Benzyloxy-5-bromopyrimidine
C11H9BrN2O
MFCD07375140

Specifications

742058-39-5
265.11
100° to 103°C
2-Benzyloxy-5-bromopyrimidine
C11H9BrN2O
MFCD07375140
1g
95%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning