Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Benzyloxy-5-bromopyridine, 95%

$119.92 - $270.92

Chemical Identifiers

Chemical Name or Material 2-Benzyloxy-5-bromopyridine
CAS 83664-33-9
Molecular Formula C12H10BrNO
Formula Weight 264.12
MDL Number MFCD09037462
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2756103

 
alfa aesar™
H2756103
1g Each for $270.92

AAH27561MD

 
alfa aesar™
H27561MD
250mg Each for $119.92
Description & Specifications

Specifications

Formula Weight 264.12
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-Benzyloxy-5-bromopyridine
Assay 95%
Molecular Formula C12H10BrNO
MDL Number MFCD09037462
CAS 83664-33-9
Melting Point 45° to 48°C
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.