Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Benzyloxy-3-hydroxypyridine, 95%

$95.12 - $215.50

Chemical Identifiers

Chemical Name or Material 2-Benzyloxy-3-hydroxypyridine
CAS 885952-26-1
Molecular Formula C12H11NO2
Formula Weight 201.22
MDL Number MFCD06798282
Synonym 2-Benzyloxy-3-pyridinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2736403

 
alfa aesar™
H2736403
1g Each for $215.50

AAH27364MD

 
alfa aesar™
H27364MD
250mg Each for $95.12
Description & Specifications

Specifications

Formula Weight 201.22
Chemical Name or Material 2-Benzyloxy-3-hydroxypyridine
Assay 95%
Molecular Formula C12H11NO2
MDL Number MFCD06798282
CAS 885952-26-1
Synonym 2-Benzyloxy-3-pyridinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 70° to 72°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.