Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Benzyloxy-3-bromopyridine, 95%

$133.30 - $300.16

Chemical Identifiers

Chemical Name or Material 2-Benzyloxy-3-bromopyridine
CAS 52200-49-4
Molecular Formula C12H10BrNO
Formula Weight 264.12
MDL Number MFCD09037467
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2782803

 
alfa aesar™
H2782803
1g Each for $300.16

AAH27828MD

 
alfa aesar™
H27828MD
250mg Each for $133.30
Description & Specifications

Specifications

Formula Weight 264.12
Chemical Name or Material 2-Benzyloxy-3-bromopyridine
Assay 95%
Molecular Formula C12H10BrNO
MDL Number MFCD09037467
CAS 52200-49-4
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.