Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Benzylidenecyclohexanone, 98%

$86.19 - $331.49

Chemical Identifiers

Chemical Name or Material 2-Benzylidenecyclohexanone
CAS 5682-83-7
Molecular Formula C13H14O
Formula Weight 186.25
MDL Number MFCD00070483
Beilstein 638548
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1343403

  SDS
alfa aesar™
L1343403
1g Each for $86.19

AAL1343406

  SDS
alfa aesar™
L1343406
5g Each for $331.49
Description & Specifications

Specifications

Formula Weight 186.25
Chemical Name or Material 2-Benzylidenecyclohexanone
MDL Number MFCD00070483
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C13H14O
CAS 5682-83-7
Assay 98%
Quantity 1g
Health Hazard 1 Warning
Boiling Point 180° to 184°C (14mmHg)
Beilstein 638548
Melting Point 53° to 55°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.