Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Benzylidene-1-tetralone, 97%

$77.91 - $271.22

Chemical Identifiers

Chemical Name or Material 2-Benzylidene-1-tetralone
CAS 6261-32-1
Molecular Formula C17H14O
Formula Weight 234.3
MDL Number MFCD00019675
Beilstein 476209
Synonym 2-Benzylidene-1,2,3,4-tetrahydronaphthalen-1-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2469903

  SDS
alfa aesar™
B2469903
1g Each for $77.91

AAB2469906

  SDS
alfa aesar™
B2469906
5g Each for $271.22
Description & Specifications

Specifications

Formula Weight 234.3
Chemical Name or Material 2-Benzylidene-1-tetralone
Melting Point 103° to 105°C
Assay 97%
Quantity 1g
Molecular Formula C17H14O
MDL Number MFCD00019675
CAS 6261-32-1
Beilstein 476209
Synonym 2-Benzylidene-1,2,3,4-tetrahydronaphthalen-1-one