Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Benzylamino-4-benzyloxy-5-bromopyrimidine, 95%

$127.76 - $289.38

Chemical Identifiers

Chemical Name or Material 2-Benzylamino-4-benzyloxy-5-bromopyrimidine
CAS 885952-25-0
Molecular Formula C18H16BrN3O
Formula Weight 370.25
MDL Number MFCD06798242
Synonym N-Benzyl-4-benzyloxy-5-bromo-2-pyrimidinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2737203

  SDS
alfa aesar™
H2737203
1g Each for $289.38

AAH27372MD

  SDS
alfa aesar™
H27372MD
250mg Each for $127.76
Description & Specifications

Specifications

Formula Weight 370.25
Chemical Name or Material 2-Benzylamino-4-benzyloxy-5-bromopyrimidine
Assay 95%
Molecular Formula C18H16BrN3O
MDL Number MFCD06798242
CAS 885952-25-0
Synonym N-Benzyl-4-benzyloxy-5-bromo-2-pyrimidinamine
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.