Please login to your online account to display your discounted pricing

2-(Benzylamino)-1,3-thiazole-5-carboxylic acid, ≥95%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-(Benzylamino)-1, 3-thiazole-5-carboxylic acid
CAS 342405-23-6
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C11H10N2O2S
Formula Weight 234.28
MDL Number MFCD03086200
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO00042CB

 
fisher scientific company
MO00042CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 234.28
Quantity 250mg
Assay 95%
CAS Min % 95.0
Chemical Name or Material 2-(Benzylamino)-1, 3-thiazole-5-carboxylic acid
Molecular Formula C11H10N2O2S
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 95%
MDL Number MFCD03086200
CAS 342405-23-6
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.