Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Benzyl-4-chlorophenol, 96%

Chemical Identifiers

Chemical Name or Material 2-Benzyl-4-chlorophenol
CAS 120-32-1
Molecular Formula C13H11ClO
Formula Weight 218.69
MDL Number MFCD00020140
UN Number UN3077
Beilstein 1959194
Merck Index 14,2407
Synonym Clorophene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2011236

  SDS
alfa aesar™
B2011236
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2011214

  SDS
alfa aesar™
B2011214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2011222

  SDS
alfa aesar™
B2011222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-Benzyl-4-chlorophenol
CAS 120-32-1
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number GO7175000
TSCA TSCA
Assay 96%
Molecular Formula C13H11ClO
Formula Weight 218.69
MDL Number MFCD00020140
UN Number UN3077
Beilstein 1959194
Merck Index 14,2407
Synonym Clorophene
Boiling Point 160° to 162°C (5mmHg)
Melting Point 45° to 51°C
Quantity 500g
Flash Point 185°C (365°F)

Applications
2-Benzyl-4-chlorophenol is used in formulating disinfectant and sanitizer products. It is also used in soaps, anionic detergents, cosmetics and aerosol spray products. It is a useful biochemical for proteomics research.

Solubility
Soluble in ethanol, alkaline solutions. Soluble in water(<1mg/mL).

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.