Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Benzyl-4,4-dimethyl-2-oxazoline, 95%

$181.64 - $181.64

Chemical Identifiers

Chemical Name or Material 2-Benzyl-4,4-dimethyl-2-oxazoline
CAS 1569-08-0
Molecular Formula C12H15NO
Formula Weight 189.26
MDL Number MFCD00102140
Synonym 2-Benzyl-4,4-dimethyl-4,5-dihydrooxazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2514703

 
alfa aesar™
B2514703
1g Each for $181.64
Description & Specifications

Specifications

Formula Weight 189.26
Chemical Name or Material 2-Benzyl-4,4-dimethyl-2-oxazoline
Assay 95%
Molecular Formula C12H15NO
MDL Number MFCD00102140
CAS 1569-08-0
Synonym 2-Benzyl-4,4-dimethyl-4,5-dihydrooxazole
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number RP6798960
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.