missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Benzyl-1,3-dioxolane 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC388680250

 View more versions of this product

Catalog No. AC388680250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥92%
Liquid
Clear colorless to pale yellow
92.0
Glass bottle
2-Benzyl-1, 3-dioxolane
94%
1.5200 to 1.5220 (20°C, 589nm)
164.2
25g
1.085g/mL
100.0
Authentic
MFCD00003215
C10H12O2
101-49-5

Chemical Identifiers

101-49-5
100.0
C10H12O2
MFCD00003215
≥92%
92.0
94%
164.2
2-Benzyl-1, 3-dioxolane

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.