missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Benzyl-1,3-dioxolane 98%, ACROS Organics™

Chemical Identifiers

CAS 101-49-5
CAS Min % 92.0
CAS Max % 100.0
Assay Percent Range 94%
Molecular Formula C10H12O2
Formula Weight 164.2
MDL Number MFCD00003215
Chemical Name or Material 2-Benzyl-1, 3-dioxolane
Assay ≥92%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC388680250 SDS Thermo Fisher Scientific
AC388680250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC388681000 SDS Thermo Fisher Scientific
AC388681000
Glass bottle 100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

101-49-5
100.0
C10H12O2
MFCD00003215
≥92%
92.0
94%
164.2
2-Benzyl-1, 3-dioxolane

Specifications

101-49-5
100.0
Glass bottle
1.085g/mL
Liquid
25g
164.2
2-Benzyl-1, 3-dioxolane
92.0
Authentic
1.5200 to 1.5220 (20°C, 589nm)
Clear colorless to pale yellow
94%
C10H12O2
MFCD00003215
≥92%

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.