missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Benzoylthiophene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1203609
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Benzoylthiophene | |
55°C to 57°C | |
C11H8OS | |
10 g | |
2-benzoylthiophene, phenyl thiophen-2-yl methanone, phenyl 2-thienyl ketone, methanone, phenyl-2-thienyl, ketone, phenyl 2-thienyl, phenyl 2-thienyl methanone, 2-thienyl phenyl ketone, .alpha.-benzoylthiophene, phenyl-thiophen-2-yl-methanone, 2-benzoyl thiophene | |
C1=CC=C(C=C1)C(=O)C2=CC=CS2 | |
188.244 | |
188.25 |
135-00-2 | |
124°C to 126°C (0.5 mmHg) | |
MFCD00022495 | |
3929 | |
DWYFUJJWTRPARQ-UHFFFAOYSA-N | |
phenyl(thiophen-2-yl)methanone | |
67262 | |
98% |
Chemical Identifiers
135-00-2 | |
188.244 | |
DWYFUJJWTRPARQ-UHFFFAOYSA-N | |
67262 | |
C1=CC=C(C=C1)C(=O)C2=CC=CS2 |
C11H8OS | |
MFCD00022495 | |
2-benzoylthiophene, phenyl thiophen-2-yl methanone, phenyl 2-thienyl ketone, methanone, phenyl-2-thienyl, ketone, phenyl 2-thienyl, phenyl 2-thienyl methanone, 2-thienyl phenyl ketone, .alpha.-benzoylthiophene, phenyl-thiophen-2-yl-methanone, 2-benzoyl thiophene | |
phenyl(thiophen-2-yl)methanone |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 205-169-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only