missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Benzoylacetanilide, TCI America™

$58.06 - $148.20

Chemical Identifiers

CAS 959-66-0
Molecular Formula C15H13NO2
Molecular Weight (g/mol) 239.274
MDL Number MFCD00003084
InChI Key XRZDIHADHZSFBB-UHFFFAOYSA-N
Synonym 2-benzoylacetanilide, benzoylacetanilide, 3-oxo-3-phenylpropionanilide, benzenepropanamide, .beta.-oxo-n-phenyl, acetamide,n-2-benzoylphenyl, 3-oxo-3-phenyl-n-phenylpropanamide, alpha-benzoylacetanilide, benzoylacetoanilide, .alpha.-benzoylacetanilide, .alpha.-benzoylacetoanilide
PubChem CID 70398
IUPAC Name 3-oxo-N,3-diphenylpropanamide
SMILES C1=CC=C(C=C1)C(=O)CC(=O)NC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B09045G View Documents TCI America
B09045G
5g Each for $58.06
 
B090425G View Documents TCI America
B090425G
25g Each for $148.20
 
Specifications

Chemical Identifiers

959-66-0
239.274
XRZDIHADHZSFBB-UHFFFAOYSA-N
70398
C1=CC=C(C=C1)C(=O)CC(=O)NC2=CC=CC=C2
C15H13NO2
MFCD00003084
2-benzoylacetanilide, benzoylacetanilide, 3-oxo-3-phenylpropionanilide, benzenepropanamide, .beta.-oxo-n-phenyl, acetamide,n-2-benzoylphenyl, 3-oxo-3-phenyl-n-phenylpropanamide, alpha-benzoylacetanilide, benzoylacetoanilide, .alpha.-benzoylacetanilide, .alpha.-benzoylacetoanilide
3-oxo-N,3-diphenylpropanamide

Specifications

White-Yellow
5g
C15H13NO2
2-benzoylacetanilide, benzoylacetanilide, 3-oxo-3-phenylpropionanilide, benzenepropanamide, .beta.-oxo-n-phenyl, acetamide,n-2-benzoylphenyl, 3-oxo-3-phenyl-n-phenylpropanamide, alpha-benzoylacetanilide, benzoylacetoanilide, .alpha.-benzoylacetanilide, .alpha.-benzoylacetoanilide
C1=CC=C(C=C1)C(=O)CC(=O)NC2=CC=CC=C2
239.274
239.27
≥98.0% (N)
107°C
959-66-0
MFCD00003084
XRZDIHADHZSFBB-UHFFFAOYSA-N
3-oxo-N,3-diphenylpropanamide
70398
Crystal-Powder at 20°C
2-Benzoylacetanilide
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1388

TSCA : No