missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Benzoxazolinone, 98%, ACROS Organics™

Manufacturer:  Acros Organics 169081000

 View more versions of this product

Catalog No. AC169081000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Benzoxazolinone
Glass bottle
137°C to 141°C
59-49-4
100.0
C7H5NO2
2-benzoxazolinone, benzoxazolinone, benzoxazolone, 2-hydroxybenzoxazole, 2-benzoxazolol, 2 3h-benzoxazolone, 1,3-benzoxazol-2 3h-one, 2-benzoxazolone, benzo d oxazol-2 3h-one, benzoxazolin-2-one
C1=CC=C2C(=C1)NC(=O)O2
135.122
135.12
≥97.5%
Authentic
Beige to Brown-Gray
100g
97.5
98%
MFCD00005716
ASSKVPFEZFQQNQ-UHFFFAOYSA-N
3H-1,3-benzoxazol-2-one
6043
Powder

Chemical Identifiers

59-49-4
135.122
ASSKVPFEZFQQNQ-UHFFFAOYSA-N
6043
C1=CC=C2C(=C1)NC(=O)O2
C7H5NO2
MFCD00005716
2-benzoxazolinone, benzoxazolinone, benzoxazolone, 2-hydroxybenzoxazole, 2-benzoxazolol, 2 3h-benzoxazolone, 1,3-benzoxazol-2 3h-one, 2-benzoxazolone, benzo d oxazol-2 3h-one, benzoxazolin-2-one
3H-1,3-benzoxazol-2-one
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents