missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Benzothiazoleacetonitrile, 98%

Manufacturer:  Alfa Aesar™ A1134314

 View more versions of this product

Catalog No. AAA1134314


 

Description

Description

Applications
2-Benzothiazoleacetonitrile is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dye stuffs.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents .
Specifications

Specifications

2-Benzothiazoleacetonitrile
98%
MFCD00051633
128460
99°C to 103°C
C1=CC=C2C(=C1)N=C(S2)CC#N
174.221
342465
56278-50-3
C9H6N2S
UN3439
2-Cyanomethyl-1,3-benzothiazole; 2-(Cyanomethyl)benzothiazole
ZMZSYUSDGRJZNT-UHFFFAOYSA-N
2-(1,3-benzothiazol-2-yl)acetonitrile
25g
174.23

Chemical Identifiers

56278-50-3
174.221
ZMZSYUSDGRJZNT-UHFFFAOYSA-N
342465
C9H6N2S
MFCD00051633
2-Cyanomethyl-1,3-benzothiazole; 2-(Cyanomethyl)benzothiazole
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

SDS Documents