Please login to your online account to display your discounted pricing

2-Benzo[b]furan-3-ylethylamine, ≥97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Benzo[b]furan-3-ylethylamine
CAS 27404-31-5
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H11NO
Formula Weight 161.2
MDL Number MFCD08271905
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC40813DA

  SDS
fisher scientific company
CC40813DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 161.2
Quantity 1g
Assay 97%
CAS Max % 100.0
Packaging Amber Glass Bottle
Chemical Name or Material 2-Benzo[b]furan-3-ylethylamine
MDL Number MFCD08271905
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 27404-31-5
CAS Min % 96.5
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula C10H11NO
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Assay Percent Range 97%