Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Benzimidazolylthio)acetic acid hydrazide, 97%

$107.43 - $406.82

Chemical Identifiers

Chemical Name or Material 2-(Benzimidazolylthio)acetic acid hydrazide
CAS 30065-27-1
Molecular Formula C9H10N4O4S
Formula Weight 222.27
MDL Number MFCD00171027
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1719806

  SDS
alfa aesar™
A1719806
5g Each for $107.43

AAA1719814

  SDS
alfa aesar™
A1719814
25g Each for $406.82
Description & Specifications

Specifications

Formula Weight 222.27
MDL Number MFCD00171027
Molecular Formula C9H10N4O4S
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-(Benzimidazolylthio)acetic acid hydrazide
CAS 30065-27-1
Quantity 5g
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈160°C (dec.)
Assay 97%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.